4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane) 99

Supplier: BeanTown Chemical

Synonyms: 4-Nitrobenzo-15-crown-5

142440-1G 142440-5G
BT142440-1GEA 50 USD
BT142440-1G BT142440-5G
4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane) 99
4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane)

CAS: 60835-69-0; MDL No: MFCD00060713 Granular; Molecular Formula: C14H19NO7; MW: 313.30 Melting Point: 93-97°


  • Granular

Formula: C₁₄H₁₉NO₇
MW: 313.30 g/mol
Melting Pt: 93…97 °C
MDL Number: MFCD00060713
CAS Number: 60835-69-0

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR