4-Cyano-3-fluorophenyl-4-n-pentylbenzoate ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-Pentylbenzoic acid 4-Cyano-3-fluorophenyl ester, 4-Amylbenzoic acid 4-Cyano-3-fluorophenyl ester, 4-Cyano-3-fluorophenyl 4-Pentylbenzoate, 4-Cyano-3-fluorophenyl 4-Amylbenzoate

142380-1G 142380-5G
BT142380-1GEA 103 USD
BT142380-1G BT142380-5G
4-Cyano-3-fluorophenyl-4-n-pentylbenzoate ≥97%
4-Cyano-3-fluorophenyl-4-n-pentylbenzoate

CAS: 86786-89-2; MDL No: MFCD11053363 Solid; Molecular Formula: C19H18FNO2; MW: 311.36 Melting Point: 29-32°


  • Solid

Formula: C₁₉H₁₈FNO₂
MW: 311.356 g/mol
Melting Pt: 29…32 °C
MDL Number: MFCD11053363
CAS Number: 86786-89-2

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR