Bis(pentamethylcyclopentadienyl)iron(II) ≥99%
Supplier: BeanTown Chemical
Synonyms:
Bis(pentamethylcyclopentadienyl)iron, Decamethylferrocene
142350-1G
142350-250MG
BT142350-1GEA
201.58
USD
BT142350-1G
BT142350-250MG
Bis(pentamethylcyclopentadienyl)iron(II) ≥99%
Bis(pentamethylcyclopentadienyl)iron (II)
CAS: 12126-50-0; MDL No: MFCD00058736
Crystalline; Molecular Formula: C20H30Fe ; MW: 326.31
Melting Point: 277° (decomposes)
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Crystalline
Formula:
C₂₀H₃₀Fe MW: 326.30 g/mol Melting Pt: 277 °C (dec.) |
MDL Number:
MFCD00058736 CAS Number: 12126-50-0 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...