4-Cyanobenzaldehyde ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Formylbenzonitrile

142315-100G 142315-25G 142315-5G
BT142315-100GEA 572 USD
BT142315-100G BT142315-25G BT142315-5G
4-Cyanobenzaldehyde ≥98%
4-Cyanobenzaldehyde

CAS: 105-07-7; EC No: 203-267-3; MDL No: MFCD00003376; RTECS: CU5250000 Powder; Molecular Formula: C8H5NO; MW: 131.13 Melting Point: 100-102°; Boiling Point: 133°/12 mmHg Air Sensitive


  • RTECS: CU5250000
  • Air Sensitive
  • Powder

Formula: C₈H₅NO
MW: 131.13 g/mol
Boiling Pt: 133 °C/12 mmHg
Melting Pt: 100…102 °C
MDL Number: MFCD00003376
CAS Number: 105-07-7
EINECS: 203-267-3
UN: 3439
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR