p-Anisaldehyde ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-Methoxybenzaldehyde
142310-1KG
142310-250G
142310-50G
BT142310-1KGEA
101.1
USD
BT142310-1KG
BT142310-250G
BT142310-50G
p-Anisaldehyde ≥98%
p-Anisaldehyde
CAS: 123-11-5; EC No: 204-602-6; MDL No: MFCD00003385; RTECS: BZ2625000
Liquid; Molecular Formula: C8H8O2; MW: 136.15
Melting Point: -1°; Boiling Point: 248°
Density (g/mL): 1.119; Refractive Index: 1.573
Air Sensitive, Light Sensitive
- RTECS: BZ2625000
- Refractive index: 1.573
- Air Sensitive, Light Sensitive
- Liquid
Formula:
C₈H₈O₂ Boiling Pt: 248 °C Melting Pt: –1 °C Density: 1.119 g/mL |
MDL Number:
MFCD00003385 CAS Number: 123-11-5 EINECS: 204-602-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...