Boron nitride ≥99.5%

Supplier: BeanTown Chemical

142300-250G 142300-50G
BT142300-250GEA 374.36 USD
BT142300-250G BT142300-50G
Boron nitride ≥99.5%
Boron nitride

CAS: 10043-11-5; EC No: 233-136-6; MDL No: MFCD00011317 10 Micron Powder; Molecular Formula: BN; MW: 24.82


  • 10 Micron Powder

Formula: BN
MW: 24.82 g/mol
Melting Pt: 2527
MDL Number: MFCD00011317
CAS Number: 10043-11-5
EINECS: 233-136-6
Merck Index: 13,01336

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR