Silicon nitride ≥99.9% (trace metals basis) 90% min α-phase

Supplier: BeanTown Chemical

Synonyms: Silicon(IV) nitride

142290-100G 142290-25G 142290-500G
BT142290-100GEA 146 USD
BT142290-100G BT142290-25G BT142290-500G
Silicon nitride ≥99.9% (trace metals basis) 90% min α-phase
Silicon nitride

CAS: 12033-89-5; EC No: 234-796-8; MDL No: MFCD00011230; RTECS: VW0650000 1 Micron Powder; Linear Formula: Si3N4; MW: 140.28 Melting Point: 1900° (sublimes) Density (g/mL): 3.44


  • RTECS: VW0650000
  • <1 Micron Powder

Formula: N₄Si₃
MW: 140.28 g/mol
Melting Pt: 1900 °C (subl.)
Density: 3.44 g/mL
MDL Number: MFCD00011230
CAS Number: 12033-89-5

Order Now

Specification Test Results

Assay 99.9%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR