Dexamethasone ≥98%
Supplier: BeanTown Chemical
Synonyms:
Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
142255-1G
142255-25G
142255-5G
BT142255-1GEA
127.1
USD
BT142255-1G
BT142255-25G
BT142255-5G
Dexamethasone ≥98%
Dexamethasone
CAS: 50-02-2; EC No: 200-003-9; MDL No: MFCD00064136; RTECS: TU3980000
Powder; Molecular Formula: C22H29FO5; MW: 392.46
Melting Point: 262-264°
Light Sensitive
- RTECS: TU3980000
- Light Sensitive
- Powder
Formula:
C₂₂H₂₉FO₅ MW: 392.47 g/mol Melting Pt: 262…264 °C |
MDL Number:
MFCD00064136 CAS Number: 50-02-2 EINECS: 200-003-9 |
Specification Test Results
Assay | 98% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...