5-Bromo-2-nitropyridine ≥98%
Supplier: BeanTown Chemical
142230-1G
142230-5G
BT142230-1GEA
37
USD
BT142230-1G
BT142230-5G
5-Bromo-2-nitropyridine ≥98%
5-Bromo-2-nitropyridine
CAS: 39856-50-3; MDL No: MFCD00160411
Solid; Molecular Formula: C6H3BrN2O2 ; MW: 202.99
Melting Point: 148-150°
- Solid
Formula:
C₅H₃BrN₂O₂ Melting Pt: 148…150 °C |
MDL Number:
MFCD00160411 CAS Number: 39856-50-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...