Potassium-2-ethylhexanoate hydrate ≥95%

Supplier: BeanTown Chemical

142195-25G 142195-5G
BT142195-25GEA 269 USD
BT142195-25G BT142195-5G
Potassium-2-ethylhexanoate hydrate ≥95%
Potassium-2-ethylhexanoate hydrate

CAS: 3164-85-0; EC No: 221-625-7; MDL No: MFCD00045896 Lump/Semiliquid; Molecular Formula: C8H15KO2·xH2O ; MW: 182.30 (anhydrous) Moisture Sensitive


  • Lump/Semiliquid
  • Moisture Sensitive

Formula: C₈H₁₅KO₂·xH₂O
MDL Number: MFCD00149471
CAS Number: 3164-85-0
EINECS: 221-625-7

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR