4-Methyl-2-hexylamine hydrochloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Methylhexanamine Hydrochloride, 2-Amino-4-methylhexane Hydrochloride, 1,3-Dimethylamylamine Hydrochloride, 1,3-Dimethylpentylamine Hydrochloride

142180-10MG 142180-50MG
BT142180-10MGEA 62 USD
BT142180-10MG BT142180-50MG
4-Methyl-2-hexylamine hydrochloride ≥98%
4-Methyl-2-hexylamine hydrochloride

CAS: 13803-74-2; MDL No: MFCD00025613 Solid; Linear Formula: CH3[CH2CH(CH3)]2NH2·HCl; Molecular Formula: C7H17N·HCl; MW: 151.68


  • Solid

Formula: C₇H₁₇N
MW: 151.68 g/mol
MDL Number: MFCD09038354
CAS Number: 13803-74-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR