1,3-Dibutylurea ≥98%
Supplier: BeanTown Chemical
Synonyms:
N,N'-Dibutylurea, N,N'-Di-n-butylurea
142165-25G
142165-5G
BT142165-25GEA
189
USD
BT142165-25G
BT142165-5G
1,3-Dibutylurea ≥98%
1,3-Dibutylurea
CAS: 1792-17-2; EC No: 217-258-7; MDL No: MFCD00015229
Solid; Linear Formula: CH3(CH2)3NHCONH(CH2)3CH3; Molecular Formula: C9H20N2O; MW: 172.27
Melting Point: 69-75°
- Solid
Formula:
H₃CCH₂CH₂CH₂NHC(O)NHCH₂CH₂CH₂CH₃ MW: 172.27 g/mol Melting Pt: 69…75 °C |
MDL Number:
MFCD00015229 CAS Number: 1792-17-2 EINECS: 217-258-7 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...