7-Bromo-1,2,3,4-tetrahydroisoquinoline ≥97%
Supplier: BeanTown Chemical
142130-1G
142130-5G
BT142130-1GEA
92
USD
BT142130-1G
BT142130-5G
7-Bromo-1,2,3,4-tetrahydroisoquinoline ≥97%
7-Bromo-1,2,3,4-tetrahydroisoquinoline
CAS: 17680-55-6; MDL No: MFCD06739047
Solid; Molecular Formula: C9H10BrN; MW: 212.09
Melting Point: 32-35°
- Solid
Formula:
C₉H₁₀BrN Melting Pt: 32…35 °C |
MDL Number:
MFCD06739047 CAS Number: 17680-55-6 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...