Chroman-4-one ≥98%

Supplier: BeanTown Chemical

142125-100G 142125-25G 142125-5G
BT142125-100GEA 586 USD
BT142125-100G BT142125-25G BT142125-5G
Chroman-4-one ≥98%
Chroman-4-one

CAS: 491-37-2; EC No: 207-736-3; MDL No: MFCD00006840; RTECS: DJ2981225 Solid; Molecular Formula: C9H8O2; MW: 148.16 Melting Point: 36-40°; Boiling Point: 127-128°; Flash point: 110°C (230°F) Refractive Index: 1.575


  • RTECS: DJ2981225
  • Refractive index: 1.575
  • Solid

Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 127…128 °C
Melting Pt: 36…40 °C
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00006840
CAS Number: 491-37-2
EINECS: 207-736-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR