Sodium diphenylamine-4-sulphonate ≥98%

Supplier: BeanTown Chemical

Synonyms: Sodium Diphenylaminesulphonate, Sodium N-phenylsulphanilate, Sodium (4-anilinobenzenesulphonate), Diphenylamine-4-sulphonic acid, sodium salt

142110-25G 142110-5G
BT142110-25GEA 269 USD
BT142110-25G BT142110-5G
Sodium diphenylamine-4-sulphonate ≥98%
Sodium diphenylamine-4-sulphonate

CAS: 6152-67-6; MDL No: MFCD00036468 Powder; Molecular Formula: C12H10NNaO3S ; MW: 271.27


  • Powder

Formula: C₁₂H₁₀NNaO₃S
MDL Number: MFCD00036468
CAS Number: 6152-67-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR