tert-Butyl (4R-cis)-6-formaldehyde-2,2-dimethyl-1,3-dioxane-4-acetate
Supplier: BeanTown Chemical
Synonyms:
tert-Butyl (3R,5S)-6-oxo-3,5-o-iso-propylidene-3,5-dihydroxyhexanoate, tert-Butyl (4R-cis)-6-formaldehydel-2,2-dimethyl-1,3-dioxane-4-acetate, (4R-cis)-6-Formaldehyde-2,2-dimethyl-1,3-dioxane-4-acetic acid 1,1-dimethylethyl ester
142085-1G
142085-5G
BT142085-1GEA
135
USD
BT142085-1G
BT142085-5G
tert-Butyl (4R-cis)-6-formaldehyde-2,2-dimethyl-1,3-dioxane-4-acetate
tert-Butyl (4R-cis)-6-formaldehyde-2,2-dimethyl-1,3-dioxane-4-acetate
CAS: 124752-23-4
Solid; Molecular Formula: C13H22O5; MW: 258.31
Formula:
C₁₃H₂₂O₅ |
MDL Number:
MFCD02093270 CAS Number: 124752-23-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...