8-Methylquinoline ≥97%

Supplier: BeanTown Chemical

142065-25G 142065-5G
BT142065-25GEA 128 USD
BT142065-25G BT142065-5G
8-Methylquinoline ≥97%
8-Methylquinoline

CAS: 611-32-5; EC No: 210-264-0; MDL No: MFCD00006810; RTECS: VC0562000 Liquid; Molecular Formula: C10H9N; MW: 143.19 Melting Point: -80°; Boiling Point: 143°/34 mmHg; Flash point: 105°C (221°F) Density (g/mL): 1.052; Refractive Index: 1.614


  • RTECS: VC0562000
  • Refractive index: 1.614
  • Liquid

Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 143 °C/34 mmHg
Melting Pt: –80 °C
Density: 1.052 g/mL
Flash Pt: 105 °C (221 °F)
MDL Number: MFCD00006810
CAS Number: 611-32-5
EINECS: 210-264-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR