(±)-Mandelic acid ≥99%

Supplier: BeanTown Chemical

Synonyms: DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid

142050-100G 142050-2.5KG 142050-500G
BT142050-100GEA 27.52 USD
BT142050-100G BT142050-2.5KG BT142050-500G
(±)-Mandelic acid ≥99%
(±)-Mandelic acid

CAS: 90-64-2; EC No: 202-007-6; MDL No: MFCD00064250; RTECS: OO6300000 Solid; Linear Formula: C6H5CH(OH)CO2H; Molecular Formula: C8H8O3; MW: 152.15 Melting Point: 119-121° Light Sensitive


  • RTECS: OO6300000
  • Light Sensitive
  • Solid

Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 119…121 °C
MDL Number: MFCD00064250
CAS Number: 90-64-2
EINECS: 202-007-6

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR