Polysorbate 40

Supplier: BeanTown Chemical

142045-100ML 142045-500ML
BT142045-100MLEA 23 USD
BT142045-100ML BT142045-500ML
Polysorbate 40
Polysorbate 40

CAS: 9005-66-7; MDL No: MFCD00165345; RTECS: WG2933000 Liquid Boiling Point: 100°; Flash point: 110°C (230°F) Density (g/mL): 1.083; Refractive Index: 1.470


  • RTECS: WG2933000
  • Liquid
  • Refractive Index: 1.470

Formula: C₆₂H₁₂₂O₂₆
Boiling Pt: >100 °C
Density: 1.083 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00165345
CAS Number: 9005-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR