3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥98%
Supplier: BeanTown Chemical
Synonyms:
Thiazolyl blue tetrazolium bromide, MTT, 2-(4,5-Dimethylthiazol-2-yl)-3,5-diphenyl-2H-tetrazol-3-ium bromide
142015-1G
142015-5G
BT142015-1GEA
46
USD
BT142015-1G
BT142015-5G
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥98%
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)
CAS: 298-93-1; EC No: 206-069-5; MDL No: MFCD00011964; RTECS: XF8060000
Crystalline/Powder; Molecular Formula: C18H16BrN5S; MW: 414.33
Melting Point: 195° (decomposes)
Air Sensitive, Light Sensitive, Moisture Sensitive
- RTECS: XF8060000
- Air Sensitive, Light Sensitive, Moisture Sensitive
- Crystalline/Powder
Formula:
C₁₈H₁₆N₅BrS MW: 414.33 g/mol Melting Pt: 195 °C (dec.) |
MDL Number:
MFCD00011964 CAS Number: 298-93-1 EINECS: 206-069-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...