1,2,4-Triazole ≥99%

Supplier: BeanTown Chemical

Synonyms: 1,2,4-1H-Triazole, 1H-1,2,4-Triazole

141950-100G 141950-25G 141950-500G
BT141950-100GEA 120 USD
BT141950-100G BT141950-25G BT141950-500G
1,2,4-Triazole ≥99%
1,2,4-Triazole

CAS: 288-88-0; EC No: 206-022-9; MDL No: MFCD00005228; RTECS: XZ3806000 Crystalline/Flakes; Molecular Formula: C2H3N3 ; MW: 69.07 Melting Point: 119-121°; Boiling Point: 260°; Flash point: 170°C (338°F)


  • RTECS: XZ3806000
  • Crystalline/Flakes

Formula: C₂H₃N₃
MW: 69.07 g/mol
Boiling Pt: 260 °C
Melting Pt: 119…121 °C
Flash Pt: 170 °C (338 °F)
MDL Number: MFCD00005228
CAS Number: 288-88-0
EINECS: 206-022-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR