2-Amino-4,6-dichloropyrimidine-5-carbaldehyde ≥96%

Supplier: BeanTown Chemical

Synonyms: 2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde

141915-1G 141915-5G
BT141915-1GEA 99 USD
BT141915-1G BT141915-5G
2-Amino-4,6-dichloropyrimidine-5-carbaldehyde ≥96%
2-Amino-4,6-dichloropyrimidine-5-carbaldehyde

CAS: 5604-46-6; MDL No: MFCD03001242 Solid; Molecular Formula: C5H3Cl2N3O; MW: 192.00 Melting Point: 208-224° Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₅H₃Cl₂N₃O
MW: 192.00 g/mol
Melting Pt: 208…224 °C
MDL Number: MFCD03001242
CAS Number: 5604-46-6

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR