3-tert-Butylaniline ≥97%

Supplier: BeanTown Chemical

Synonyms: 3-(tert-Butyl)aniline, m-tert-Butylaniline

141910-1G 141910-5G
BT141910-1GEA 187 USD
BT141910-1G BT141910-5G
3-tert-Butylaniline ≥97%
3-tert-Butylaniline

CAS: 5369-19-7; EC No: 226-361-6; MDL No: MFCD00125078 Liquid; Molecular Formula: C10H15N; MW: 149.23


  • Liquid

Formula: C₁₀H₁₅N
MW: 149.24 g/mol
MDL Number: MFCD00125078
CAS Number: 5369-19-7
EINECS: 226-361-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR