Dilithium metasilicate ≥99.5% (trace metals basis)
Supplier: BeanTown Chemical
141905-100G
141905-500G
BT141905-100GEA
184.72
USD
BT141905-100G
BT141905-500G
Dilithium metasilicate ≥99.5% (trace metals basis)
Dilithium metasilicate
CAS: 10102-24-6; EC No: 233-270-5; MDL No: MFCD00016179
Powder; Linear Formula: Li2SO3; MW: 89.94
- Powder
Formula:
Li₂O₃Si MW: 89.97 g/mol |
MDL Number:
MFCD00016179 CAS Number: 10102-24-6 EINECS: 233-270-5 |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...