(+)-Di-p-toluoyl-D-tartaric acid ≥98%

Supplier: BeanTown Chemical

Synonyms: Di-p-toluoyl-D-tartaric acid, (2S,3S)-(+)-Di-O-4-Toluoyl-D-Tartaric Acid, Di-p-toluoyl-D-tartaric Acid

141870-100G 141870-25G 141870-5G
BT141870-100GEA 273 USD
BT141870-100G BT141870-25G BT141870-5G
(+)-Di-p-toluoyl-D-tartaric acid ≥98%
(+)-Di-p-toluoyl-D-tartaric acid

CAS: 32634-68-7; EC No: 251-132-2; MDL No: MFCD00008552 Powder; Molecular Formula: C20H18O8 ; MW: 386.35 Melting Point: 169-171° Optical Rotation: [α]19/D +138°, c = 1 in ethanol


  • Optical rotation: [α]19/D +138°, c = 1 in ethanol
  • Powder

Formula: C₂₀H₁₈O₈
Melting Pt: 169…171 °C
MDL Number: MFCD00008552
CAS Number: 32634-68-7
EINECS: 251-132-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR