Calcium chloride, anhydrous ≥93%

Supplier: BeanTown Chemical

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

141865-1KG 141865-5KG
BT141865-1KGEA 61.15 USD
BT141865-1KG BT141865-5KG
Calcium chloride, anhydrous ≥93%
Calcium chloride

CAS: 10043-52-4; EC No: 233-140-8; MDL No: MFCD00010903; RTECS: EV9800000 Powder/Granular; Molecular Formula: CaCl2; MW: 110.98 Melting Point: 772° Hygroscopic


  • RTECS: EV9800000
  • Hygroscopic
  • Powder/Granular

Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
Merck Index: 14,01659

Order Now

Specification Test Results

Assay 93%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR