2-Amino-3-pyridinemethanol ≥98%

Supplier: BeanTown Chemical

Synonyms: (2-Amino-pyridin-3-yl)-methanol, 3-(Hydroxymethyl)-2-pyridinamine, 2-Amino-3-pyridinemethanol, 2-Amino-3-hydroxymethylpyridine, 2-Amino-3-pyridylmethanol, 2-Amino-3-(hydroxymethyl)pyridine, 2-Aminopyridine-3-methanol

141835-1G 141835-5G
BT141835-1GEA 174 USD
BT141835-1G BT141835-5G
2-Amino-3-pyridinemethanol ≥98%
2-Amino-3-pyridinemethanol

CAS: 23612-57-9; MDL No: MFCD05663510 Solid; Molecular Formula: C6H8N2O ; MW: 124.14 Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₆H₈N₂O
MW: 124.14 g/mol
MDL Number: MFCD05663510
CAS Number: 23612-57-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR