2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Chloro-5-(trifluoromethyl)isonicotinic acid
141805-1G
141805-250MG
141805-5G
BT141805-1GEA
76
USD
BT141805-1G
BT141805-250MG
BT141805-5G
2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid ≥98%
2-Chloro-5-(trifluoromethyl)pyridine-4-carboxylic acid
CAS: 505084-58-2
Solid; Molecular Formula: C7H3ClF3NO2; MW: 225.55
- Solid
Formula:
C₇H₃ClF₃NO₂ MW: 225.55 g/mol |
MDL Number:
MFCD01862019 CAS Number: 505084-58-2 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...