2,6-Di-tert-butyl-4-methylpyridine ≥97%
Supplier: BeanTown Chemical
141720-1G
141720-5G
BT141720-1GEA
35
USD
BT141720-1G
BT141720-5G
2,6-Di-tert-butyl-4-methylpyridine ≥97%
2,6-Di-tert-butyl-4-methylpyridine
CAS: 38222-83-2; EC No: 253-834-4; MDL No: MFCD00006305
Powder; Molecular Formula: C14H23N; MW: 205.34
Melting Point: 33-36°; Boiling Point: 233°; Flash point: 84°C (183°F)
Refractive Index: 1.476
Air Sensitive, Light Sensitive
- Refractive index: 1.476
- Air Sensitive, Light Sensitive
- Powder
Formula:
C₁₄H₂₃N Boiling Pt: 233 °C Melting Pt: 33…36 °C Flash Pt: 84 °C (183 °F) |
MDL Number:
MFCD00006305 CAS Number: 38222-83-2 EINECS: 253-834-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...