Rhenium pentachloride ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
141695-1G
141695-5G
BT141695-1GEA
183
USD
BT141695-1G
BT141695-5G
Rhenium pentachloride ≥99.9% (trace metals basis)
Rhenium pentachloride
CAS: 13596-35-5; EC No: 237-042-6; MDL No: MFCD00011197
-50 Mesh Powder; Linear Formula: ReCl5; MW: 363.47
Moisture Sensitive
- Moisture Sensitive
- -50 Mesh Powder
Formula:
Cl₅Re |
MDL Number:
MFCD00011197 CAS Number: 13596-35-5 EINECS: 237-042-6 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...