1,3,5-Trimethoxybenzene ≥99%

Supplier: BeanTown Chemical

Synonyms: O,O,O-1,3,5-Trimethylresorcinol

141625-10G 141625-250G 141625-50G
BT141625-10GEA 40.75 USD
BT141625-10G BT141625-250G BT141625-50G
1,3,5-Trimethoxybenzene ≥99%
1,3,5-Trimethoxybenzene

CAS: 621-23-8; EC No: 210-673-4; MDL No: MFCD00008385; RTECS: DC2810000 Crystalline; Molecular Formula: C9H12O3 ; MW: 168.19 Melting Point: 50-53°; Boiling Point: 255°; Flash point: 86°C (187°F)


  • RTECS: DC2810000
  • Crystalline

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 255 °C
Melting Pt: 50…53 °C
Flash Pt: 86 °C (187 °F)
MDL Number: MFCD00008385
CAS Number: 621-23-8
EINECS: 210-673-4

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR