Nickel (II) oxide ≥99.995% (trace metals basis)
Supplier: BeanTown Chemical
Synonyms:
Nickel monoxide, Nickel oxide, Bunsenite
141620-100G
141620-25G
141620-5G
BT141620-100GEA
413.36
USD
BT141620-100G
BT141620-25G
BT141620-5G
Nickel (II) oxide ≥99.995% (trace metals basis)
Nickel (II) oxide
CAS: 1313-99-1; EC No: 215-215-7; MDL No: MFCD00011145; RTECS: QR8400000
Powder; Molecular Formula: NiO; MW: 74.71
Melting Point: 1984°
Density (g/mL): 6.67
- RTECS: QR8400000
- Powder
Formula:
NiO MW: 74.69 g/mol Melting Pt: 1984 °C Density: 6.67 g/mL |
MDL Number:
MFCD00011145 CAS Number: 1313-99-1 EINECS: 215-215-7 |
Specification Test Results
Assay | 99.995% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...