6-Quinolinol ≥95%
Supplier: BeanTown Chemical
141610-25G
141610-5G
BT141610-25GEA
172
USD
BT141610-25G
BT141610-5G
6-Quinolinol ≥95%
6-Quinolinol
CAS: 580-16-5; EC No: 209-454-6; MDL No: MFCD00047611; RTECS: VC4130000
Powder; Molecular Formula: C9H7NO; MW: 145.16
Melting Point: 188-190°
Light Sensitive
- RTECS: VC4130000
- Light Sensitive
- Powder
Formula:
C₉H₇NO Melting Pt: 188…190 °C |
MDL Number:
MFCD00047611 CAS Number: 580-16-5 EINECS: 209-454-6 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...