Ammonium thiocyanate ≥97.5% ACS

Supplier: BeanTown Chemical

Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate

141590-1KG 141590-250G
BT141590-1KGEA 204.88 USD
BT141590-1KG BT141590-250G
Ammonium thiocyanate ≥97.5% ACS
Ammonium thiocyanate

CAS: 1762-95-4; EC No: 217-175-6; MDL No: MFCD00011428; RTECS: XK7875000 Crystalline; Linear Formula: NH4SCN; MW: 76.12 Melting Point: 152-154° Hygroscopic , Light Sensitive


  • RTECS: XK7875000
  • Hygroscopic, Light Sensitive
  • Crystalline

Formula: NH₄SCN
MW: 76.12 g/mol
Melting Pt: 150
MDL Number: MFCD00011428
CAS Number: 1762-95-4
EINECS: 217-175-6
Merck Index: 13,00564

Order Now

Specification Test Results

Assay 97.5% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR