Isopropyl-4-hydroxybenzoate ≥98%
Supplier: BeanTown Chemical
141580-10G
141580-250G
141580-50G
BT141580-10GEA
46
USD
BT141580-10G
BT141580-250G
BT141580-50G
Isopropyl-4-hydroxybenzoate ≥98%
Isopropyl-4-hydroxybenzoate
CAS: 4191-73-5; EC No: 224-069-3; MDL No: MFCD00016468; RTECS: DH2250000
Solid; Molecular Formula: C10H12O3 ; MW: 180.2
Melting Point: 84-86°
- RTECS: DH2250000
- Solid
Formula:
C₁₀H₁₂O₃ Melting Pt: 84…86 °C |
MDL Number:
MFCD00016468 CAS Number: 4191-73-5 EINECS: 224-069-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...