1,6-Dibromohexane ≥97%

Supplier: BeanTown Chemical

141545-100G 141545-2.5KG 141545-500G
BT141545-100GEA 39.14 USD
BT141545-100G BT141545-2.5KG BT141545-500G
1,6-Dibromohexane ≥97%
1,6-Dibromohexane

CAS: 629-03-8; EC No: 211-067-2; MDL No: MFCD00000272; RTECS: MO1515000 Liquid; Linear Formula: Br(CH2)6Br ; MW: 243.97 Melting Point: -2°; Boiling Point: 243°; Flash point: 110°C (230°F) Density (g/mL): 1.586; Refractive Index: 1.507


  • RTECS: MO1515000
  • Refractive index: 1.507
  • Liquid

Formula: BrH₂C(CH₂)₄CH₂Br
Boiling Pt: 243 °C
Melting Pt: –2 °C
Density: 1.586 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00000272
CAS Number: 629-03-8
EINECS: 211-067-2
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR