D-(-)-Aspartic acid ≥99%

Supplier: BeanTown Chemical

141535-100G 141535-25G 141535-5G
BT141535-100GEA 260 USD
BT141535-100G BT141535-25G BT141535-5G
D-(-)-Aspartic acid ≥99%
D-(-)-Aspartic acid

CAS: 1783-96-6; EC No: 217-234-6; MDL No: MFCD00063081; RTECS: CI9097500 Powder; Molecular Formula: C8H12N2O8; MW: 133.10 Melting Point: 300° Density (g/mL): 1.66; Optical Rotation: [α]20/D -24°, c = 2.3 in 6M HCl


  • RTECS: CI9097500
  • Optical rotation: [α]20/D -24°, c = 2.3 in 6M HCl
  • Powder

Formula: C₄H₇NO₄
Melting Pt: >300 °C
Density: 1.66 g/mL
MDL Number: MFCD00063081
CAS Number: 1783-96-6
EINECS: 217-234-6

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR