9-Fluorenylmethyl carbamate ≥98%

Supplier: BeanTown Chemical

Synonyms: Carbamic Acid 9-Fluorenylmethyl Ester, Fmoc-NH2, (9H-Fluoren-9-ylmethoxy)carboxamide

141520-1G 141520-5G
BT141520-1GEA 106 USD
BT141520-1G BT141520-5G
9-Fluorenylmethyl carbamate ≥98%
9-Fluorenylmethyl carbamate

CAS: 84418-43-9; MDL No: MFCD00237376 Powder; Molecular Formula: C15H13NO2; MW: 239.27 Melting Point: 201-204°


  • Powder

Formula: C₁₅H₁₃NO₂
MW: 239.27 g/mol
Melting Pt: 201…204 °C
MDL Number: MFCD00237376
CAS Number: 84418-43-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR