5-Bromopicolinic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 5-Bromopyridine-2-carboxylic acid, 5-Bromo-2-pyridinecarboxylic acid

141495-1G 141495-25G 141495-5G
BT141495-1GEA 46 USD
BT141495-1G BT141495-25G BT141495-5G
5-Bromopicolinic acid ≥98%
5-Bromopicolinic acid

CAS: 30766-11-1; MDL No: MFCD00234149 Powder; Molecular Formula: C6H4B2NO2; MW: 202.01 Melting Point: 173-176°


  • Powder

Formula: C₆H₄BrNO₂
MW: 202.01 g/mol
Melting Pt: 173…176 °C
MDL Number: MFCD00234149
CAS Number: 30766-11-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR