Strontium hexafluoro-2,4-pentanedionate

Supplier: BeanTown Chemical

Synonyms: Strontium hexafluoroacetylacetonate

141485-1G 141485-5G
BT141485-1GEA 37 USD
BT141485-1G BT141485-5G
Strontium hexafluoro-2,4-pentanedionate
Strontium hexafluoro-2,4-pentanedionate

CAS: 121012-89-3; MDL No: MFCD00064760 Crystalline; Molecular Formula: C10H2F12O4Sr; MW: 501.75 Melting Point: 260° (decomposes); Boiling Point: 220°/0.2 mmHg


  • Crystalline

Formula: C₁₀H₂F₁₂O₄Sr
MW: 501.75 g/mol
Boiling Pt: 220 °C/0.2 mmHg
Melting Pt: 260 °C (dec.)
MDL Number: MFCD00064760
CAS Number: 121012-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR