(±)-Camphorquinone ≥99%

Supplier: BeanTown Chemical

Synonyms: 1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, DL-Camphoroquinone

141480-100G 141480-25G 141480-5G
BT141480-100GEA 571 USD
BT141480-100G BT141480-25G BT141480-5G
(±)-Camphorquinone ≥99%
(±)-Camphorquinone

CAS: 10373-78-1; EC No: 233-814-1; MDL No: MFCD00064160 Powder; Molecular Formula: C10H14O2; MW: 166.22 Melting Point: 197-203°


  • Powder

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
Melting Pt: 197…203 °C
MDL Number: MFCD00064160
CAS Number: 10373-78-1
EINECS: 233-814-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR