Adenosine-5'-triphosphate disodium salt (ATP disodium salt) ≥98%

Supplier: BeanTown Chemical

Synonyms: ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Adenosine 5'-triphosphoric acid disodium, Adenosine 5′-triphosphate disodium salt, ATP 2Na, ATP disodium salt, Adenosine 5ʹ-Triphosphate, Disodium Salt

141470-1G 141470-5G
BT141470-1GEA 40.89 USD
BT141470-1G BT141470-5G
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) ≥98%
Adenosine 5'-triphosphate disodium salt (ATP disodium salt)

CAS: 987-65-5; EC No: 213-579-1; MDL No: MFCD00150755; RTECS: AU7417000 Powder; Molecular Formula: C10H14N5Na2O13P3; MW: 551.14 Melting Point: 176° (decomposes)


  • RTECS: AU7417000
  • Powder

Formula: C₁₀H₁₄N₅Na₂O₁₃P₃
MW: 551.14 g/mol
Melting Pt: 176 °C (dec.)
MDL Number: MFCD00080339
CAS Number: 987-65-5
EINECS: 213-579-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR