Bis(triphenylphosphoranylidene)ammonium chloride, (max. 3% H₂O) ≥97% (dry weight)
Supplier: BeanTown Chemical
Synonyms:
1,1,1-Triphenyl-N-(triphenylphosphoranylidene)phosphoraniminium chloride, Bis(triphenylphosphine)iminium chloride
141410-25G
141410-5G
BT141410-25GEA
250
USD
BT141410-25G
BT141410-5G
Bis(triphenylphosphoranylidene)ammonium chloride, (max. 3% H₂O) ≥97% (dry weight)
Bis(triphenylphosphoranylidene)ammonium chloride
CAS: 21050-13-5; EC No: 244-170-6; MDL No: MFCD00151523
Powder; Molecular Formula: C36H30ClNP2; MW: 574.03
Melting Point: 270-272°
- Powder
Formula:
C₃₆H₃₀ClNP₂ MW: 574.041 g/mol Melting Pt: 270…272 °C |
MDL Number:
MFCD00151523 CAS Number: 21050-13-5 EINECS: 244-170-6 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...