Tetraoctylammonium bromide ≥98%
Supplier: BeanTown Chemical
Synonyms:
Tetra-n-octylammonium bromide
141370-10G
141370-250G
141370-50G
BT141370-10GEA
56.21
USD
BT141370-10G
BT141370-250G
BT141370-50G
Tetraoctylammonium bromide ≥98%
Tetraoctylammonium bromide
CAS: 14866-33-2; EC No: 238-936-9; MDL No: MFCD00011863
Crystalline; Linear Formula: [CH3(CH2)7]4N(Br); Molecular Formula: C32H68BrN; MW: 546.79
Melting Point: 95-98°
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
C₃₂H₆₈BrN MW: 546.8 g/mol Melting Pt: 95…98 °C |
MDL Number:
MFCD00011863 CAS Number: 14866-33-2 EINECS: 238-936-9 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...