Fmoc-Dap(Boc)-OH ≥95%

Supplier: BeanTown Chemical

Synonyms: Fmoc-(N-beta-boc)-L-alpha,beta-diaminopropionic acid, 3-(tert-Butoxycarbonyl)amino-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-alanine, (S)-3-(tert-Butoxycarbonylamino)-2-[(9H-fluoren-9-ylmethoxy)carbonylamino]propionic acid, Nα-Fmoc-Nβ-Boc-L-2,3-diaminopropionic acid, (S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 3-(Boc-amino)-N-Fmoc-L-alanine

141330-1G 141330-250MG 141330-5G
BT141330-1GEA 121 USD
BT141330-1G BT141330-250MG BT141330-5G
Fmoc-Dap(Boc)-OH ≥95%
Fmoc-Dap(Boc)-OH

CAS: 162558-25-0; MDL No: MFCD00235896 Powder; Molecular Formula: C23H26N2O6; MW: 426.46 Melting Point: 131-139°


  • Powder

Formula: C₂₃H₂₆N₂O₆
MW: 426.47 g/mol
Melting Pt: 131…139 °C
MDL Number: MFCD00235896
CAS Number: 162558-25-0

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR