D-(+)-Xylose ≥98%
Supplier: BeanTown Chemical
141315-100G
141315-2.5KG
141315-500G
BT141315-100GEA
40.89
USD
BT141315-100G
BT141315-2.5KG
BT141315-500G
D-(+)-Xylose ≥98%
D-(+)-Xylose
CAS: 58-86-6; EC No: 200-400-7; MDL No: MFCD00151475; RTECS: ZF2285000
Crystalline; Molecular Formula: C5H10O5; MW: 150.13
Melting Point: 154-158°
Hygroscopic
- RTECS: ZF2285000
- Hygroscopic
- Crystalline
Formula:
C₅H₁₀O₅ Melting Pt: 154…158 °C |
MDL Number:
MFCD00151475 CAS Number: 58-86-6 EINECS: 200-400-7 |
|
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...