Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99%

Supplier: BeanTown Chemical

141280-100G 141280-2.5KG 141280-500G
BT141280-100GEA 27.98 USD
BT141280-100G BT141280-2.5KG BT141280-500G
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99%
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

CAS: 77-86-1; EC No: 201-064-4; MDL No: MFCD00004679; RTECS: TY2900000 Crystalline; Linear Formula: NH2C(CH2OH)3; Molecular Formula: C4H11NO3; MW: 121.14 Melting Point: 169°; Boiling Point: 288° (decomposes below boiling point) Density (g/mL): 1.353 Hygroscopic


  • RTECS: TY2900000
  • Hygroscopic
  • Crystalline

Formula: NH₂C(CH₂OH)₃
Boiling Pt: 288 °C (dec.)
Melting Pt: 169 °C
Density: 1.353 g/mL
MDL Number: MFCD00004679
CAS Number: 77-86-1
EINECS: 201-064-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR