Tin(II) sulphate ≥95%

Supplier: BeanTown Chemical

Synonyms: Tin(II) sulphate, Tin sulphate, Stannous sulphate

141255-1KG 141255-250G
BT141255-1KGEA 173 USD
BT141255-1KG BT141255-250G
Tin(II) sulphate ≥95%
Tin(II) sulphate

CAS: 7488-55-3; EC No: 231-302-2; MDL No: MFCD00011246 Powder; Linear Formula: SnSO4; MW: 214.77 Melting Point: 378° (decomposes) Density (g/mL): 4.15 Moisture Sensitive


  • Moisture Sensitive
  • Powder

Formula: O₄SSn
MW: 214.77 g/mol
Melting Pt: 378 °C (dec.)
Density: 4.15 g/mL
MDL Number: MFCD00011246
CAS Number: 7488-55-3
EINECS: 231-302-2
Merck Index: 12,08947

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR