Dimethyl maleate ≥96%

Supplier: BeanTown Chemical

Synonyms: Dimethyl Maleate, Maleic acid dimethyl ester

141245-1KG 141245-250G
BT141245-1KGEA 114 USD
BT141245-1KG BT141245-250G
Dimethyl maleate ≥96%
Dimethyl maleate

CAS: 624-48-6; EC No: 210-848-5; MDL No: MFCD00008459; RTECS: EM6300000 Liquid; Linear Formula: CH3OCOCH=CHCOOCH3; Molecular Formula: C6H8O4; MW: 144.13 Boiling Point: 204-205°; Flash point: 95°C (203°F) Density (g/mL): 1.152; Refractive Index: 1.441


  • RTECS: EM6300000
  • Refractive index: 1.441
  • Liquid

Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 205
Melting Pt: -17.5
Density: 1.152 g/mL
Flash Pt: 95 °C (203 °F)
MDL Number: MFCD00008459
CAS Number: 624-48-6
EINECS: 210-848-5

Order Now

Specification Test Results

Assay 96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR