2-(2-Hydroxyethyl)-1-methylpyrrolidine ≥98%

Supplier: BeanTown Chemical

Synonyms: 1-Methyl-2-pyrrolidineethanol

141235-25G 141235-5G
BT141235-25GEA 249 USD
BT141235-25G BT141235-5G
2-(2-Hydroxyethyl)-1-methylpyrrolidine ≥98%
2-(2-Hydroxyethyl)-1-methylpyrrolidine

CAS: 67004-64-2; EC No: 266-538-5; MDL No: MFCD00003174 Liquid; Molecular Formula: C7H15NO; MW: 129.20 Boiling Point: 110-112°/14 mmHg; Flash point: 84°C (183°F) Density (g/mL): 0.951; Refractive Index: 1.471


  • Refractive index: 1.471
  • Liquid

Formula: C₇H₁₅NO
MW: 129.20 g/mol
Boiling Pt: 110…112 °C/14 mmHg
Density: 0.951 g/mL
Flash Pt: 84 °C (183 °F)
MDL Number: MFCD00003174
CAS Number: 67004-64-2
EINECS: 266-538-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR